Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143984.1 | NP_001137456.1 | 1 | 538 | ||
NM_175732.2 | NP_783859.1 | 2 | 429 | |||
GenBank mRNA | AF277187.1 | - | 1 | 35 | ||
AK294640.1 | - | 2 | 260 | |||
AY333987.1 | - | 2 | 236 | |||
BC014048.1 | - | 1 | 75 | |||
BC020242.1 | - | 2 | 258 | 4591438 | ||
BC107573.1 | - | 1 | 373 | |||
DA476984.1 | - | 2 | 285 | |||
DR004030.1 | - | 2 | 287 |