Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191013.1 | NP_001177942.1 | 3, 4 | 769 | ||
NM_001191014.1 | NP_001177943.1 | 1, 2 | 204 | |||
NM_001191015.1 | NP_001177944.1 | 1, 2 | 204 | |||
NM_183239.1 | NP_899062.1 | 3, 4 | 769 | |||
XM_006717624.3 | XP_006717687.1 | 3, 4 | 458 | |||
XM_011539270.2 | XP_011537572.1 | 3, 4 | 1832 | |||
XM_011539271.1 | XP_011537573.1 | 3, 4 | 458 | |||
XM_011539272.2 | XP_011537574.1 | 3, 4 | 458 | |||
XM_011539273.2 | XP_011537575.1 | 3, 4 | 458 | |||
XM_017015670.1 | XP_016871159.1 | 3, 4 | 458 | |||
XM_017015671.1 | XP_016871160.1 | 3, 4 | 458 | |||
GenBank mRNA | AK291886.1 | - | 3, 4 | 457 | ||
AK296266.1 | - | 2, 3 | 156 | |||
AK297522.1 | - | 1, 2 | 167 | |||
AK300508.1 | - | 2, 3 | 166 | |||
AY191318.1 | - | 2, 3 | 137 | |||
AY209189.1 | - | 3, 4 | 461 | |||
AY350731.1 | - | 3, 4 | 461 | |||
BC023583.2 | - | 1, 2 | 170 | |||
BC056918.1 | - | 3, 4 | 769 | 5165084 | ||
BM708208.1 | - | 1, 2 | 108 | |||
DC341647.1 | - | 2, 3 | 156 |