Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144601.3 | NP_653202.1 | 5 | 747 | ||
NM_181553.2 | NP_853531.1 | 4 | 583 | |||
NR_037613.1 | 5 | 967 | ||||
NR_037619.1 | 4 | 640 | ||||
XM_006721131.1 | XP_006721194.1 | 5 | 791 | |||
XM_011522861.1 | XP_011521163.1 | 3 | 420 | |||
GenBank mRNA | AF479813.1 | - | 5 | 721 | ||
AK056324.1 | - | 5 | 967 | |||
AK075075.1 | - | 3 | 280 | |||
BC023509.1 | - | 4 | 489 | 3960441 | ||
BC023591.2 | - | 4 | 481 | 4638047 | ||
BC036042.1 | - | 4 | 543 | 5271808 | ||
BC040227.1 | - | 4 | 534 | 5187826 | ||
BC047701.1 | - | 4 | 583 | 5770132 | ||
BI457147.1 | - | 4 | 435 | |||
CT001840.1 | - | 4 | 630 |