Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152345.4 | NP_689558.4 | 12 | 1409 | ||
XM_005257920.4 | XP_005257977.1 | 12 | 1375 | |||
XM_011524319.2 | XP_011522621.1 | 12 | 1612 | |||
XM_017024174.1 | XP_016879663.1 | 12 | 1309 | |||
XM_017024175.1 | XP_016879664.1 | 12 | 1311 | |||
XM_017024176.1 | XP_016879665.1 | 12 | 1241 | |||
GenBank mRNA | AK092673.1 | - | 12 | 1226 | ||
AK097716.1 | - | 11 | 1245 | |||
AK098421.1 | - | 4 | 365 | |||
BC032554.1 | - | 12 | 1456 | 5557374 | ||
BC062597.1 | - | 11 | 1259 | |||
BG029526.1 | - | 190 |