Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098817.1 | NP_001092287.1 | 5 | 591 | ||
NM_001308064.1 | NP_001294993.1 | 3 | 354 | |||
NM_194281.3 | NP_919257.2 | 3 | 483 | |||
XM_005258196.4 | XP_005258253.1 | 3 | 301 | |||
XM_005258197.4 | XP_005258254.1 | 3 | 514 | |||
XM_011525812.2 | XP_011524114.1 | 3 | 349 | |||
XM_017025549.1 | XP_016881038.1 | 3 | 321 | |||
GenBank mRNA | AK095502.1 | - | 3 | 425 | ||
AK300660.1 | - | 5 | 543 | |||
BC039404.1 | - | 3 | 449 | 5300795 | ||
BC048989.1 | - | 4 | 411 | |||
BM758571.1 | - | 3 | 256 | |||
BQ441485.1 | - | 3 | 357 | |||
BQ887287.1 | - | 3 | 483 | |||
BY800049.1 | - | 5 | 506 |