Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102597.2 | NP_001096067.2 | 5 | 815 | ||
NM_001102598.2 | NP_001096068.2 | 5 | 815 | |||
NM_001102599.2 | NP_001096069.2 | 5 | 815 | |||
NM_001102600.2 | NP_001096070.2 | 5 | 815 | |||
XM_011526429.2 | XP_011524731.1 | 6 | 1469 | |||
XM_011526430.2 | XP_011524732.1 | 6 | 1470 | |||
XM_011526431.2 | XP_011524733.1 | 6 | 1471 | |||
XM_011526432.2 | XP_011524734.1 | 2 | 133 | |||
GenBank mRNA | AY358129.1 | - | 5 | 1078 | ||
EU008600.1 | - | 5 | 815 | |||
EU008601.1 | - | 5 | 815 | |||
EU008602.1 | - | 5 | 815 | |||
EU008603.1 | - | 5 | 815 |