Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271830.1 | NP_001258759.1 | 2 | 80 | ||
NM_174895.2 | NP_777555.1 | 2 | 362 | |||
XM_006722639.2 | XP_006722702.1 | 2 | 582 | |||
XM_017026249.1 | XP_016881738.1 | 4 | 1391 | |||
XM_017026250.1 | XP_016881739.1 | 4 | 1250 | |||
XM_017026251.1 | XP_016881740.1 | 3 | 1206 | |||
XM_017026252.1 | XP_016881741.1 | 2 | 723 | |||
GenBank mRNA | AK130855.1 | - | 2 | 314 | ||
BC025387.1 | - | 2 | 367 | 4824135 | ||
BC038715.1 | - | 2 | 298 | 5268414 | ||
BX281028.1 | - | 2 | 39 | |||
BX647746.1 | - | 2 | 110 |