Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001299.5 | NP_001290.2 | 2, 3 | 387 | ||
NM_001308341.1 | NP_001295270.1 | 3, 4 | 662 | |||
NM_001308342.1 | NP_001295271.1 | 3, 4 | 653 | |||
XM_005259741.2 | XP_005259798.1 | 2, 3 | 236 | |||
XM_017026289.1 | XP_016881778.1 | 2, 3 | 559 | |||
GenBank mRNA | AK223234.1 | - | 2, 3 | 253 | ||
AK300837.1 | - | 2, 3 | 167 | |||
AK301888.1 | - | 3, 4 | 526 | |||
AK313255.1 | - | 2, 3 | 256 | |||
AK316406.1 | - | 3, 4 | 603 | |||
BC022015.2 | - | 2, 3 | 340 | 4824832 | ||
BC036307.1 | - | 2, 3 | 326 | 5165618 | ||
BG718473.1 | - | 2, 3 | 390 | |||
D17408.1 | - | 2, 3 | 268 | |||
EU794601.1 | - | 2, 3 | 326 | |||
S80560.1 | - | 2, 3 | 250 | |||
U37019.1 | - | 2, 3 | 254 |