Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162916.1 | NP_001156388.1 | 4 | 494 | ||
NM_138467.2 | NP_612476.1 | 5 | 589 | |||
NR_027962.1 | 5 | 680 | ||||
XM_006710347.1 | XP_006710410.1 | 5 | 602 | |||
GenBank mRNA | AK098237.1 | - | 5 | 589 | ||
AK294264.1 | - | 2 | 319 | |||
AK299849.1 | - | 5 | 445 | |||
AK311272.1 | - | 5 | 626 | |||
BC009514.1 | - | 5 | 523 | |||
BC071765.1 | - | 5 | 589 | 3931600 | ||
BX647591.1 | - | 1 | 36 |