Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195736.1 | NP_001182665.1 | 2, 3 | 397 | ||
NM_001195737.1 | NP_001182666.1 | 2, 3 | 397 | |||
NM_001195738.1 | NP_001182667.1 | 2, 3 | 397 | |||
NM_001195740.1 | NP_001182669.1 | 2, 3 | 397 | |||
NM_001195741.1 | NP_001182670.1 | 2, 3 | 397 | |||
NM_152371.3 | NP_689584.2 | 2, 3 | 397 | |||
NR_036637.1 | 2, 3 | 330 | ||||
NR_036638.1 | 2, 3 | 330 | ||||
XM_006710354.3 | XP_006710417.1 | 2, 3 | 365 | |||
XM_011540664.1 | XP_011538966.1 | 2, 3 | 367 | |||
XM_011540665.1 | XP_011538967.1 | 2, 3 | 367 | |||
XM_011540666.1 | XP_011538968.1 | 2, 3 | 367 | |||
GenBank mRNA | AF425266.1 | - | 2, 3 | 341 | ||
AK057027.1 | - | 2, 3 | 328 | |||
AK075273.1 | - | 2, 3 | 349 | |||
AK094901.1 | - | 1, 2 | 211 | |||
AK291908.1 | - | 2, 3 | 355 | |||
AK298926.1 | - | 2, 3 | 351 | |||
AK303504.1 | - | 2, 3 | 369 | |||
AK308034.1 | - | 2, 3 | 307 | |||
AK316243.1 | - | 2, 3 | 307 | |||
BC022547.1 | - | 2, 3 | 337 | 4812314 | ||
BI908668.1 | - | 2, 3 | 249 | |||
BQ709693.1 | - | 233 | ||||
DA292832.1 | - | 2, 3 | 329 | |||
DA495138.1 | - | 2, 3 | 330 | |||
DC351872.1 | - | 2, 3 | 372 | |||
DC407959.1 | - | 2, 3 | 397 |