Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172069.3 | NP_742066.2 | 27 | 4087 | ||
XM_017003351.1 | XP_016858840.1 | 26 | 4074 | |||
XM_017003353.1 | XP_016858842.1 | 28 | 4198 | |||
GenBank mRNA | AB095948.1 | - | 25 | 3873 | ||
AL832207.1 | - | 27 | 4080 | |||
AL833400.1 | - | 20 | 2390 | |||
AL833473.1 | - | 2 | 222 | |||
AL833546.1 | - | 27 | 6016 |