Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195685.1 | NP_001182614.1 | 5 | 789 | ||
NM_194317.3 | NP_919298.1 | 5 | 779 | |||
GenBank mRNA | AK124733.1 | - | 5 | 789 | ||
AK309248.1 | - | 6 | 943 | |||
AL831852.1 | - | 2 | 223 | |||
AY358623.1 | - | 5 | 695 | |||
BC032836.1 | - | 8 | 1311 | |||
BC047013.1 | - | 6 | 809 | 5171270 |