Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321300.1 | NP_001308229.1 | 5, 6 | 1448 | ||
NM_001321301.1 | NP_001308230.1 | 5, 6 | 2160 | |||
NM_001321302.1 | NP_001308231.1 | 6, 7 | 868 | |||
NM_001321305.1 | NP_001308234.1 | 4, 5 | 2044 | |||
NM_152392.4 | NP_689605.1 | 4, 5 | 2044 | |||
XM_017003377.1 | XP_016858866.1 | 5, 6 | 1002 | |||
GenBank mRNA | AF542548.1 | - | 3, 4 | 245 | ||
AK092817.1 | - | 4, 5 | 1448 | |||
AL833813.1 | - | 4, 5 | 1779 | |||
BC063789.1 | - | 4, 5 | 1933 | |||
BC113671.1 | - | 4, 5 | 1041 | 8322723 | ||
BC117278.1 | - | 4, 5 | 1041 | 40125829 |