Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301790.1 | NP_001288719.1 | 3 | 588 | ||
NM_001301791.1 | NP_001288720.1 | 4 | 723 | |||
NM_001301792.1 | NP_001288721.1 | 4 | 751 | |||
NM_152396.3 | NP_689609.2 | 4 | 723 | |||
XM_005264867.3 | XP_005264924.1 | 4 | 723 | |||
XM_005264870.4 | XP_005264927.1 | 4 | 723 | |||
XM_006712970.3 | XP_006713033.1 | 4 | 599 | |||
XM_006712972.3 | XP_006713035.1 | 4 | 723 | |||
XM_011533357.2 | XP_011531659.1 | 4 | 723 | |||
XM_017005718.1 | XP_016861207.1 | 4 | 599 | |||
XM_017005719.1 | XP_016861208.1 | 3 | 581 | |||
XM_017005720.1 | XP_016861209.1 | 3 | 581 | |||
XM_017005721.1 | XP_016861210.1 | 3 | 581 | |||
XM_017005722.1 | XP_016861211.1 | 4 | 723 | |||
XM_017005723.1 | XP_016861212.1 | 3 | 581 | |||
XM_017005724.1 | XP_016861213.1 | 4 | 723 | |||
GenBank mRNA | AK057791.1 | - | 4 | 708 | ||
AK293370.1 | - | 3 | 581 | |||
BC022400.1 | - | 4 | 680 | 4693314 | ||
BU183521.1 | - | 4 | 661 | |||
CA430635.1 | - | 3 | 453 | |||
CX872865.1 | - | 1 | 92 |