Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 12 | 1281 | ||
NM_001115008.2 | NP_001108480.1 | 12 | 1482 | |||
NM_001288996.1 | NP_001275925.1 | 9 | 1878 | |||
NM_001288997.1 | NP_001275926.1 | 12 | 1219 | |||
NM_194282.3 | NP_919258.2 | 12 | 2145 | |||
NR_110253.1 | 9 | 1804 | ||||
NR_110254.1 | 11 | 1478 | ||||
XM_005262750.4 | XP_005262807.1 | 11 | 1954 | |||
XM_006714081.3 | XP_006714144.1 | 11 | 1880 | |||
XM_017007728.1 | XP_016863217.1 | 9 | 1687 | |||
XM_017007729.1 | XP_016863218.1 | 10 | 1870 | |||
XM_017007730.1 | XP_016863219.1 | 10 | 1259 | |||
GenBank mRNA | AB111889.1 | - | 11 | 1465 | ||
AK292769.1 | - | 12 | 1229 | |||
AK294256.1 | - | 12 | 2074 | |||
AK299787.1 | - | 7 | 687 | |||
BC109277.2 | - | 9 | 1680 | |||
BC109278.2 | - | 9 | 1753 | 40028371 | ||
BX537919.1 | - | 11 | 1379 | |||
BX640657.1 | - | 12 | 1208 | |||
BX640966.1 | - | 12 | 2140 |