Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 29 | 3449 | ||
XM_011513952.1 | XP_011512254.1 | 29 | 3435 | |||
XM_011513953.1 | XP_011512255.1 | 28 | 3373 | |||
GenBank mRNA | AK097562.1 | - | 5 | 495 | ||
AK126005.1 | - | 14 | 1318 | |||
AK128183.1 | - | 19 | 1915 | |||
AL832763.1 | - | 24 | 2374 | |||
BC044937.1 | - | 17 | 1921 | |||
BC052962.1 | - | 29 | 3410 | 4838071 | ||
CR749346.1 | - | 24 | 2350 |