Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004373.3 | NP_004364.2 | 2 | 193 | ||
GenBank mRNA | AK312009.1 | - | 2 | 193 | ||
BC007723.2 | - | 1 | 158 | 4300921 | ||
BC070186.1 | - | 1 | 166 | 6654310 | ||
BC107861.1 | - | 1 | 174 | 4664958 | ||
BP378295.1 | - | 2 | 164 | |||
HE647860.1 | - | 2 | 156 | |||
X15341.1 | - | 2 | 178 |