Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286259.1 | NP_001273188.1 | 4, 5 | 1603 | ||
NM_182503.2 | NP_872309.2 | 4, 5 | 505 | |||
XM_011535439.2 | XP_011533741.1 | 7, 8 | 661 | |||
XM_011535440.1 | XP_011533742.1 | 7, 8 | 3393 | |||
XM_011535444.2 | XP_011533746.1 | 6, 7 | 524 | |||
XM_017010260.1 | XP_016865749.1 | 5, 6 | 3922 | |||
XM_017010261.1 | XP_016865750.1 | 4, 5 | 3879 | |||
XM_017010262.1 | XP_016865751.1 | 4, 5 | 383 | |||
XM_017010263.1 | XP_016865752.1 | 3, 4 | 357 | |||
GenBank mRNA | AK126201.1 | - | 4, 5 | 1609 | ||
BC037955.1 | - | 4, 5 | 465 | |||
BX538182.1 | - | 4, 5 | 903 |