Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122842.2 | NP_001116314.1 | 8 | 1089 | ||
NM_001199619.1 | NP_001186548.1 | 9 | 1072 | |||
NM_001199620.1 | NP_001186549.1 | 10 | 1190 | |||
NM_001199621.1 | NP_001186550.1 | 7 | 868 | |||
NM_181782.4 | NP_861447.3 | 8 | 1089 | |||
XM_005266822.4 | XP_005266879.1 | 8 | 1119 | |||
XM_006715340.3 | XP_006715403.1 | 8 | 1046 | |||
XM_011535455.2 | XP_011533757.1 | 8 | 994 | |||
XM_017010269.1 | XP_016865758.1 | 8 | 805 | |||
XM_017010270.1 | XP_016865759.1 | 9 | 1051 | |||
XM_017010271.1 | XP_016865760.1 | 8 | 1049 | |||
XM_017010272.1 | XP_016865761.1 | 8 | 902 | |||
XM_017010273.1 | XP_016865762.1 | 8 | 1119 | |||
XM_017010274.1 | XP_016865763.1 | 8 | 1208 | |||
GenBank mRNA | AB074156.1 | - | 7 | 617 | ||
AB074157.1 | - | 8 | 889 | |||
AF493978.1 | - | 9 | 1072 | |||
AK127512.1 | - | 8 | 899 | |||
AK294558.1 | - | 7 | 666 | |||
AK307395.1 | - | 9 | 1051 | |||
AL832621.2 | - | 8 | 882 | |||
AL832628.2 | - | 8 | 846 | |||
AL834442.1 | - | 6 | 510 | |||
BC137094.1 | - | 10 | 1106 | 9021091 | ||
BC137095.1 | - | 9 | 988 | 9021092 | ||
BX537385.1 | - | 8 | 884 |