Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPLKIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138701.3 | NP_619646.1 | 1 | 381 | ||
GenBank mRNA | AK290098.1 | - | 1 | 410 | ||
BC026265.1 | - | 1 | 346 | 4699982 | ||
BG501110.1 | - | 1 | 381 |