Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136273.1 | NP_001129745.1 | 2 | 92 | ||
XM_005274652.3 | XP_005274709.1 | 4 | 402 | |||
XM_011531115.2 | XP_011529417.1 | 3 | 353 | |||
XM_017029281.1 | XP_016884770.1 | 5 | 751 |