Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031739.2 | NP_001026909.1 | 3, 4 | 570 | ||
NM_001168530.1 | NP_001162002.1 | 3, 4 | 570 | |||
NM_001168531.1 | NP_001162003.1 | 4, 5 | 757 | |||
NM_024087.2 | NP_076992.1 | 3, 4 | 570 | |||
XM_005274446.1 | XP_005274503.1 | 4, 5 | 757 | |||
XM_017029283.1 | XP_016884772.1 | 3, 4 | 537 | |||
XM_017029285.1 | XP_016884774.1 | 3, 4 | 537 | |||
XM_017029286.1 | XP_016884775.1 | 3, 4 | 537 | |||
GenBank mRNA | AK000643.1 | - | 3, 4 | 460 | ||
AK292270.1 | - | 4, 5 | 757 | |||
AL080091.2 | - | 3, 4 | 286 | |||
BC001244.1 | - | 3, 4 | 450 | 3462727 | ||
BC013172.1 | - | 3, 4 | 520 | 3917449 |
Gene Symbol: ASB9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033769.1 | 1 | 436 |