Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199629.1 | NP_001186558.1 | 2, 3 | 496 | ||
NM_002475.4 | NP_002466.1 | 2, 3 | 432 | |||
GenBank mRNA | AK304594.1 | - | 2, 3 | 265 | ||
AK310996.1 | - | 2, 3 | 262 | |||
BC012425.1 | - | 1, 2 | 220 | 3862144 | ||
BC014400.1 | - | 2, 3 | 496 | 3503030 | ||
BU166567.1 | - | 319 | ||||
M31211.1 | - | 1, 2 | 200 | |||
X16434.1 | - | 1, 2 | 185 |