Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001125.3 | NP_001116.1 | 4 | 1136 | ||
NM_001291949.1 | NP_001278878.1 | 3 | 1050 | |||
NM_001291950.1 | NP_001278879.1 | 2 | 716 | |||
XM_011512438.2 | XP_011510740.1 | 4 | 1202 | |||
GenBank mRNA | AK291787.1 | - | 3 | 893 | ||
AK304560.1 | - | 2 | 621 | |||
AK313369.1 | - | 3 | 777 | |||
BC063883.1 | - | 4 | 893 | 30374536 | ||
BC074769.2 | - | 2 | 634 | 30915190 | ||
L13291.1 | - | 2 | 700 |