Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175058.4 | NP_778228.3 | 11 | 1531 | ||
XM_017017240.1 | XP_016872729.1 | 11 | 1538 | |||
XM_017017241.1 | XP_016872730.1 | 11 | 1538 | |||
XM_017017242.1 | XP_016872731.1 | 11 | 1538 | |||
XM_017017243.1 | XP_016872732.1 | 11 | 1538 | |||
GenBank mRNA | AK296371.1 | - | 1 | 266 | ||
AK301465.1 | - | 1 | 122 | |||
BC033239.1 | - | 2 | 408 | 5429651 | ||
BC071599.1 | - | 11 | 1527 | 30343461 |