Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282613.1 | NP_001269542.1 | 8 | 1884 | ||
NM_032735.2 | NP_116124.2 | 10 | 2146 | |||
NM_152439.3 | NP_689652.2 | 6 | 1493 | |||
XM_011537961.1 | XP_011536263.1 | 8 | 1754 | |||
XM_011537962.2 | XP_011536264.1 | 7 | 1517 | |||
XM_011537963.1 | XP_011536265.1 | 7 | 1525 | |||
XM_011537965.2 | XP_011536267.1 | 4 | 1264 | |||
GenBank mRNA | AK096459.1 | - | 10 | 2147 | ||
AY515706.1 | - | 9 | 1919 | |||
BC151138.1 | - | 8 | 1753 |