Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081492.1 | NP_001074961.1 | 4 | 703 | ||
NM_182507.2 | NP_872313.2 | 4 | 703 | |||
XM_005268676.3 | XP_005268733.1 | 2 | 798 | |||
GenBank mRNA | AJ717743.1 | - | 4 | 657 | ||
BC065180.1 | - | 4 | 703 | 6573884 |