Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152443.2 | NP_689656.2 | 5 | 619 | ||
GenBank mRNA | AK054835.1 | - | 3 | 407 | ||
AK315462.1 | - | 5 | 619 | |||
BC025724.1 | - | 3 | 465 | 5199154 | ||
KU178865.1 | - | 3 | 295 | |||
KU178866.1 | - | 3 | 295 |