Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 9, 10 | 1270 | ||
NM_001320421.1 | NP_001307350.1 | 9, 10 | 1266 | |||
XM_005267367.1 | XP_005267424.1 | 5, 6 | 581 | |||
XM_011536497.1 | XP_011534799.1 | 9, 10 | 1328 | |||
XM_017021044.1 | XP_016876533.1 | 9, 10 | 1234 | |||
XM_017021045.1 | XP_016876534.1 | 9, 10 | 1234 | |||
XM_017021046.1 | XP_016876535.1 | 9, 10 | 1328 | |||
XM_017021047.1 | XP_016876536.1 | 8, 9 | 945 | |||
GenBank mRNA | AL832848.1 | - | 8, 9 | 948 | ||
BC035865.2 | - | 3, 4 | 371 | |||
BC048270.2 | - | 3, 4 | 371 | |||
BX248275.1 | - | 5, 6 | 562 |