Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319.6 | NP_001310.3 | 4 | 786 | ||
XM_005259498.1 | XP_005259555.1 | 4 | 798 | |||
XM_005259499.3 | XP_005259556.1 | 4 | 605 | |||
XM_005259500.1 | XP_005259557.1 | 4 | 798 | |||
XM_005259501.2 | XP_005259558.1 | 4 | 1172 | |||
XM_017026297.1 | XP_016881786.1 | 4 | 672 | |||
XM_017026298.1 | XP_016881787.1 | 4 | 1172 | |||
GenBank mRNA | AB451278.1 | - | 3 | 263 | ||
AB451410.1 | - | 3 | 263 | |||
AF001177.1 | - | 4 | 1508 | |||
BC018693.2 | - | 4 | 759 | 3010717 | ||
BC018699.2 | - | 4 | 759 | 3010717 | ||
BC020972.1 | - | 3 | 316 | 3851613 | ||
BT009922.1 | - | 3 | 263 | |||
U89896.1 | - | 3 | 502 |