Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126336.2 | NP_001119808.1 | 8 | 1590 | ||
NM_001164097.1 | NP_001157569.1 | 9 | 6852 | |||
NM_001164098.1 | NP_001157570.1 | 9 | 4551 | |||
NM_004385.4 | NP_004376.2 | 10 | 9813 | |||
GenBank mRNA | AB209491.1 | - | 10 | 9696 | ||
AK296285.1 | - | 3 | 701 | |||
BX641036.1 | - | 3 | 6847 | |||
D32039.1 | - | 8 | 1339 | |||
J02814.1 | - | 3 | 1333 | |||
U16306.1 | - | 10 | 9723 | |||
U26555.1 | - | 9 | 4461 | |||
X15998.1 | - | 9 | 6762 |