Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PM20D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152491.4 | NP_689704.4 | 13 | 1502 | ||
NR_135186.1 | 12 | 1440 | ||||
XM_017000397.1 | XP_016855886.1 | 11 | 1265 | |||
XM_017000398.1 | XP_016855887.1 | 12 | 1327 | |||
GenBank mRNA | AK057131.1 | - | 12 | 1424 | ||
AK290786.1 | - | 13 | 1486 | |||
BC039170.1 | - | 13 | 1471 | 4746689 | ||
BC063477.1 | - | 13 | 1471 | 5221187 |