Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144884.1 | NP_001138356.1 | 8 | 994 | ||
NM_133496.4 | NP_598003.2 | 8 | 992 | |||
XM_011540779.2 | XP_011539081.1 | 9 | 1091 | |||
XM_017000400.1 | XP_016855889.1 | 8 | 992 | |||
XM_017000401.1 | XP_016855890.1 | 8 | 992 | |||
GenBank mRNA | AF233345.1 | - | 8 | 822 | ||
AF529197.1 | - | 8 | 807 | |||
AK313638.1 | - | 8 | 975 | |||
AY094606.1 | - | 8 | 807 | |||
BC064692.1 | - | 8 | 948 | 30330665 | ||
BX537375.1 | - | 8 | 994 | |||
CR936806.1 | - | 8 | 988 |