Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM117B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173511.3 | NP_775782.2 | 5, 6 | 1111 | ||
GenBank mRNA | AK096090.1 | - | 5, 6 | 695 | ||
BC065010.1 | - | 4, 5 | 517 | |||
BC106906.2 | - | 4, 5 | 460 | 40031780 | ||
BC106907.2 | - | 4, 5 | 456 | 40031781 |