Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 5, 6 | 798 | ||
XM_005263396.3 | XP_005263453.1 | 6, 7 | 1223 | |||
XM_011518272.1 | XP_011516574.1 | 5, 6 | 492 | |||
XM_011518273.2 | XP_011516575.1 | 4, 5 | 323 | |||
XM_011518274.1 | XP_011516576.1 | 3, 4 | 353 | |||
XM_017014301.1 | XP_016869790.1 | 5, 6 | 1079 | |||
XM_017014302.1 | XP_016869791.1 | 5, 6 | 1003 | |||
GenBank mRNA | AF116701.2 | - | 5, 6 | 777 | ||
BC130580.1 | - | 5, 6 | 457 | 40146611 | ||
BC144082.1 | - | 5, 6 | 457 | 9052602 | ||
BU949503.1 | - | 3, 4 | 360 |