Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152592.3 | NP_689805.3 | 11 | 2009 | ||
XM_005267377.3 | XP_005267434.1 | 12 | 2106 | |||
XM_006720063.3 | XP_006720126.1 | 12 | 2206 | |||
XM_011536515.2 | XP_011534817.1 | 12 | 2141 | |||
XM_017021056.1 | XP_016876545.1 | 11 | 2008 | |||
GenBank mRNA | AK098471.1 | - | 6 | 772 | ||
BX648302.1 | - | 9 | 3881 |