Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C18orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288980.1 | NP_001275909.1 | 4 | 846 | ||
NM_001288981.1 | NP_001275910.1 | 4 | 678 | |||
NM_173529.5 | NP_775800.3 | 3 | 687 | |||
XM_005258201.1 | XP_005258258.1 | 3 | 1547 | |||
XM_017025567.1 | XP_016881056.1 | 3 | 1230 | |||
GenBank mRNA | AB267906.1 | - | 3 | 1542 | ||
AB330143.1 | - | 3 | 1542 | |||
AB353076.1 | - | 4 | 845 | |||
AB353077.1 | - | 4 | 677 | |||
AK126503.1 | - | 2 | 284 | |||
BC036054.2 | - | 2 | 589 | 5273569 | ||
BX640877.1 | - | 3 | 660 |