Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148962.4 | NP_683765.1 | 1 | 883 | ||
GenBank mRNA | AB083055.2 | - | 1 | 839 | ||
BC063549.1 | - | 1 | 883 | 4777971 |