Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004398.3 | NP_004389.2 | 8 | 1190 | ||
GenBank mRNA | AB040537.1 | - | 8 | 1126 | ||
AB209884.1 | - | 8 | 1093 | |||
AK296752.1 | - | 6 | 864 | |||
AK315216.1 | - | 8 | 1121 | |||
BC049217.1 | - | 8 | 1086 | |||
BC091521.1 | - | 8 | 1138 | 30388370 | ||
BC093654.1 | - | 8 | 1094 | 7939499 | ||
BC093656.1 | - | 8 | 1094 | 7939501 | ||
U28042.1 | - | 8 | 1126 |