Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130.5 | NP_001121.2 | 2, 3 | 309 | ||
NM_198969.1 | NP_945320.1 | 2, 3 | 376 | |||
NM_198970.1 | NP_945321.1 | 2, 3 | 309 | |||
XM_006722664.1 | XP_006722727.1 | 2, 3 | 517 | |||
GenBank mRNA | AF072902.1 | - | 2, 3 | 212 | ||
AK094591.1 | - | 2, 3 | 518 | |||
AK314713.1 | - | 2, 3 | 215 | |||
BC113735.1 | - | 2, 3 | 363 | 8322787 | ||
BC113737.1 | - | 2, 3 | 363 | 8322789 | ||
BG913049.1 | - | 2, 3 | 296 | |||
BM550097.1 | - | 266 | ||||
BM908356.1 | - | 337 | ||||
U04241.1 | - | 2, 3 | 176 | |||
X73357.1 | - | 2, 3 | 130 | |||
X73358.1 | - | 1, 2 | 100 |