Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130.5 | NP_001121.2 | 6 | 486 | ||
NM_198969.1 | NP_945320.1 | 6 | 553 | |||
NM_198970.1 | NP_945321.1 | 6 | 486 | |||
XM_006722664.1 | XP_006722727.1 | 6 | 694 | |||
GenBank mRNA | AF072902.1 | - | 6 | 389 | ||
AK094591.1 | - | 6 | 695 | |||
AK095154.1 | - | 2 | 2007 | |||
AK314713.1 | - | 6 | 392 | |||
BC113735.1 | - | 6 | 540 | 8322787 | ||
BC113737.1 | - | 6 | 540 | 8322789 | ||
BG696926.1 | - | 2 | 70 | |||
BG913049.1 | - | 6 | 473 | |||
BM550097.1 | - | 443 | ||||
BM908356.1 | - | 514 | ||||
U04241.1 | - | 6 | 353 | |||
X73357.1 | - | 6 | 307 | |||
X73358.1 | - | 5 | 277 |