Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128827.2 | NP_001122299.1 | 18, 19 | 2441 | ||
NM_001321074.1 | NP_001308003.1 | 20, 21 | 3289 | |||
NM_001321075.1 | NP_001308004.1 | 18, 19 | 2450 | |||
NM_001321076.1 | NP_001308005.1 | 17, 18 | 2095 | |||
NM_001321077.1 | NP_001308006.1 | 17, 18 | 2086 | |||
NM_001365.4 | NP_001356.1 | 20, 21 | 3298 | |||
XM_005256491.1 | XP_005256548.1 | 18, 19 | 2540 | |||
XM_011523698.1 | XP_011522000.1 | 20, 21 | 3186 | |||
XM_011523699.2 | XP_011522001.1 | 21, 22 | 2428 | |||
XM_011523702.1 | XP_011522004.1 | 17, 18 | 1965 | |||
XM_017024288.1 | XP_016879777.1 | 17, 18 | 2186 | |||
XM_017024289.1 | XP_016879778.1 | 17, 18 | 2177 | |||
XM_017024290.1 | XP_016879779.1 | 17, 18 | 2655 | |||
GenBank mRNA | AK293835.1 | - | 18, 19 | 2447 | ||
AK297346.1 | - | 17, 18 | 2043 | |||
AK299801.1 | - | 17, 18 | 2057 | |||
AK308533.1 | - | 17, 18 | 2200 | |||
AK316096.1 | - | 18, 19 | 2003 | |||
BC040533.1 | - | 12, 13 | 1443 | |||
BC136553.1 | - | 20, 21 | 2822 | 9020543 | ||
U83192.1 | - | 20, 21 | 2956 |