Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286245.1 | NP_001273174.1 | 6 | 1373 | ||
NM_001286246.1 | NP_001273175.1 | 6 | 1392 | |||
NM_178502.3 | NP_848597.1 | 7 | 1553 | |||
XM_005268697.1 | XP_005268754.1 | 7 | 2056 | |||
XM_005268698.1 | XP_005268755.1 | 7 | 1829 | |||
XM_005268700.1 | XP_005268757.1 | 7 | 1709 | |||
XM_005268703.1 | XP_005268760.1 | 7 | 1728 | |||
XM_011538022.2 | XP_011536324.1 | 6 | 1931 | |||
GenBank mRNA | AK092085.1 | - | 7 | 1354 | ||
AK094385.1 | - | 6 | 1162 | |||
AK128752.1 | - | 5 | 2521 | |||
AY225126.1 | - | 4 | 1066 | |||
BC114441.1 | - | 6 | 1282 | 40083764 | ||
BC114498.1 | - | 6 | 1282 | 40083763 |