Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152649.3 | NP_689862.1 | 3, 4 | 1224 | ||
XM_005255834.3 | XP_005255891.1 | 4, 5 | 1123 | |||
XM_011522936.2 | XP_011521238.1 | 3, 4 | 1263 | |||
GenBank mRNA | AK091708.1 | - | 3, 4 | 967 |