Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195811.1 | NP_001182740.1 | 9 | 1276 | ||
NM_001195812.1 | NP_001182741.1 | 8 | 1114 | |||
NM_001320272.1 | NP_001307201.1 | 10 | 1570 | |||
NM_152666.2 | NP_689879.2 | 10 | 1563 | |||
XM_006711752.2 | XP_006711815.1 | 9 | 1730 | |||
XM_011544115.2 | XP_011542417.1 | 11 | 1638 | |||
XM_011544116.2 | XP_011542418.1 | 11 | 1466 | |||
XM_011544119.2 | XP_011542421.2 | 6 | 1637 | |||
XM_011544120.2 | XP_011542422.1 | 9 | 1282 | |||
XM_011544121.2 | XP_011542423.1 | 6 | 771 | |||
XM_011544122.2 | XP_011542424.1 | 6 | 2232 | |||
XM_017000567.1 | XP_016856056.1 | 9 | 1743 | |||
XM_017000568.1 | XP_016856057.1 | 9 | 1264 | |||
XM_017000569.1 | XP_016856058.1 | 9 | 3243 | |||
XM_017000570.1 | XP_016856059.1 | 7 | 1983 | |||
GenBank mRNA | AK091691.1 | - | 8 | 1088 | ||
AK098092.1 | - | 10 | 1570 | |||
AK295421.1 | - | 9 | 1257 | |||
AK307857.1 | - | 9 | 1255 | |||
AY391836.1 | - | 8 | 1126 | |||
AY461578.1 | - | 9 | 1276 | |||
BC101373.1 | - | 9 | 1203 | 40025860 | ||
BC101374.1 | - | 9 | 1203 | 40025861 | ||
BC101375.1 | - | 9 | 1203 | 40025862 |