Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144994.7 | NP_659431.5 | 4 | 378 | ||
GenBank mRNA | AF492401.1 | - | 4 | 354 | ||
AJ315766.1 | - | 3 | 322 | |||
AK057011.1 | - | 4 | 378 | |||
AY196212.1 | - | 4 | 362 | |||
BC107056.2 | - | 4 | 373 | 40005436 |