Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 19 | 2066 | ||
NM_004434.2 | NP_004425.2 | 18 | 2009 | |||
XM_005267397.1 | XP_005267454.1 | 19 | 2136 | |||
XM_005267398.3 | XP_005267455.1 | 19 | 2069 | |||
XM_005267399.4 | XP_005267456.2 | 19 | 2108 | |||
XM_005267400.4 | XP_005267457.1 | 18 | 2035 | |||
XM_011536540.2 | XP_011534842.1 | 18 | 2082 | |||
XM_011536541.1 | XP_011534843.1 | 19 | 2149 | |||
XM_011536542.2 | XP_011534844.1 | 18 | 1976 | |||
XM_017021074.1 | XP_016876563.1 | 17 | 1947 | |||
GenBank mRNA | AK126980.1 | - | 20 | 2022 | ||
AK299812.1 | - | 19 | 2345 | |||
BC032541.1 | - | 18 | 1985 | 5533599 | ||
BC033043.1 | - | 18 | 2009 | 4821202 | ||
BX442035.2 | - | 6 | 565 | |||
U97018.1 | - | 18 | 2008 |