Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037325.2 | NP_001032402.1 | 6, 7 | 866 | ||
NM_001199165.2 | NP_001186094.1 | 26, 27 | 2997 | |||
NM_001302891.1 | NP_001289820.1 | 24, 25 | 2947 | |||
NR_126542.1 | 5, 6 | 696 | ||||
XM_005257119.4 | XP_005257176.1 | 26, 27 | 3091 | |||
XM_005257125.3 | XP_005257182.1 | 18, 19 | 2081 | |||
XM_005257126.4 | XP_005257183.1 | 15, 16 | 1746 | |||
XM_006721740.2 | XP_006721803.1 | 26, 27 | 2963 | |||
XM_011524461.2 | XP_011522763.1 | 26, 27 | 3104 | |||
XM_011524464.2 | XP_011522766.1 | 24, 25 | 2880 | |||
XM_011524465.2 | XP_011522767.1 | 23, 24 | 2420 | |||
XM_011524466.2 | XP_011522768.1 | 16, 17 | 1899 | |||
XM_011524467.2 | XP_011522769.1 | 15, 16 | 1741 | |||
XM_017024302.1 | XP_016879791.1 | 24, 25 | 2834 | |||
GenBank mRNA | AA781351.1 | - | 1, 2 | 168 | ||
AF458591.1 | - | 16, 17 | 1904 | |||
AK098070.1 | - | 16, 17 | 1816 | |||
AK299075.1 | - | 26, 27 | 2948 | |||
BC029524.1 | - | 6, 7 | 866 | |||
BC131805.1 | - | 24, 25 | 2775 | 40119069 | ||
BG203878.1 | - | 163 | ||||
BG531260.1 | - | 3, 4 | 184 | |||
BX097629.1 | - | 6, 7 | 544 |