Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178862.2 | NP_849193.1 | 14 | 2636 | ||
XM_017005857.1 | XP_016861346.1 | 14 | 2499 | |||
XM_017005858.1 | XP_016861347.1 | 14 | 1957 | |||
XM_017005859.1 | XP_016861348.1 | 14 | 2030 | |||
GenBank mRNA | AK027789.1 | - | 4 | 520 | ||
AK075380.1 | - | 4 | 611 | |||
AX799082.1 | - | 14 | 2138 | |||
AY074880.1 | - | 14 | 2138 | |||
BC015880.1 | - | 3 | 414 |