Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 4 | 869 | ||
NM_001293214.1 | NP_001280143.1 | 3 | 653 | |||
NM_001293215.1 | NP_001280144.1 | 4 | 772 | |||
NM_001293216.1 | NP_001280145.1 | 4 | 671 | |||
NM_178014.3 | NP_821133.1 | 4 | 785 | |||
GenBank mRNA | AB062393.1 | - | 4 | 611 | ||
AF070561.1 | - | 4 | 606 | |||
AF070593.1 | - | 4 | 606 | |||
AF070600.1 | - | 4 | 623 | |||
AF141349.1 | - | 4 | 623 | |||
AK098772.1 | - | 4 | 630 | |||
AK298889.1 | - | 4 | 675 | |||
AK302314.1 | - | 4 | 738 | |||
AK303227.1 | - | 3 | 372 | |||
AK309684.1 | - | 2 | 227 | |||
AK316265.1 | - | 4 | 542 | |||
AK316316.1 | - | 4 | 667 | |||
BC001896.1 | - | 1 | 167 | |||
BC001938.1 | - | 4 | 610 | 2820585 | ||
BC002347.2 | - | 4 | 615 | 2964461 | ||
BC005838.2 | - | 4 | 610 | 2820585 | ||
BC007605.1 | - | 4 | 615 | 3350604 | ||
BC013374.2 | - | 4 | 606 | 3946253 | ||
BC019924.1 | - | 4 | 603 | 4998571 | ||
BC020946.1 | - | 4 | 644 | 4795692 | ||
BC021909.1 | - | 4 | 605 | 3509417 | ||
BC070326.1 | - | 4 | 621 | 6698679 | ||
BC103746.1 | - | 4 | 628 | 6503977 | ||
BU557299.1 | - | 4 | 785 | |||
DC389555.1 | - | 3 | 497 | |||
DQ457058.1 | - | 1 | 41 | |||
V00599.1 | - | 3 | 458 |